Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277552.1 | NP_001264481.1 | 6 - 7 | 712 | 88 | |
XM_015290626.2 | XP_015146112.1 | 7 - 8 | 761 | 88 | ||
XM_025152825.1 | XP_025008593.1 | 7 - 8 | 626 | 88 | ||
GenBank mRNA | BX929483.1 | - | 6 - 7 | 776 | 88 | |
BX929529.2 | - | 4 - 5 | 313 | 88 | ||
BX932560.1 | - | 7 - 8 | 601 | 88 | ||
BX950434.2 | - | 6 - 7 | 712 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |