Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277802.2 | NP_001264731.2 | 5 - 6 | 393 | 81 | |
NM_001277803.2 | NP_001264732.1 | 4 - 5 | 350 | 81 | ||
NM_001277804.2 | NP_001264733.1 | 4 - 5 | 453 | 81 | ||
GenBank mRNA | BU305896.1 | - | 5 - 6 | 403 | 81 | |
BU359753.1 | - | 4 - 5 | 360 | 81 | ||
BX929673.1 | - | 5 - 6 | 428 | 81 | ||
BX930584.2 | - | 4 - 5 | 372 | 81 | ||
BX933844.1 | - | 3 - 4 | 331 | 81 | ||
BX935201.2 | - | 4 - 5 | 451 | 81 | ||
BX950256.2 | - | 3 - 4 | 483 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |