Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199415.1 | NP_001186344.1 | 1 - 2 | 64 | 120 | |
GenBank mRNA | BU136820.1 | - | 1 - 2 | 67 | 120 | |
BU301058.1 | - | 1 - 2 | 44 | 120 | ||
BX929762.1 | - | 1 - 2 | 56 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |