Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199578.1 | NP_001186507.1 | 4 - 5 | 342 | 148 | |
XM_015281611.2 | XP_015137097.1 | 5 - 6 | 512 | 148 | ||
XM_015281618.2 | XP_015137104.1 | 4 - 5 | 412 | 148 | ||
XM_015281636.2 | XP_015137122.1 | 5 - 6 | 650 | 148 | ||
XM_015281639.2 | XP_015137125.1 | 5 - 6 | 475 | 148 | ||
XM_015281645.2 | XP_015137131.1 | 3 - 4 | 315 | 148 | ||
GenBank mRNA | BX930748.1 | - | 2 - 3 | 303 | 148 | |
BX933182.1 | - | 4 - 5 | 342 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |