Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277529.1 | NP_001264458.1 | 5 - 6 | 387 | 69 | |
GenBank mRNA | BX930239.1 | - | 3 - 4 | 192 | 69 | |
BX931244.1 | - | 5 - 6 | 390 | 69 | ||
BX933564.2 | - | 5 - 6 | 387 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |