Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025144866.1 | XP_025000634.1 | 3 - 4 | 580 | 116 | |
XM_025144867.1 | XP_025000635.1 | 3 - 4 | 309 | 116 | ||
XM_025144868.1 | XP_025000636.1 | 3 - 4 | 261 | 116 | ||
XM_424828.6 | XP_424828.4 | 3 - 4 | 577 | 116 | ||
GenBank mRNA | BX933855.1 | - | 1 - 2 | 50 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |