Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414580.5 | XP_414580.1 | 8 - 9 | 1036 | 77 | |
GenBank mRNA | BX934896.1 | - | 5 - 6 | 454 | 77 | |
CR385449.1 | - | 297 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |