Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414580.5 | XP_414580.1 | 10 - 11 | 1247 | 71 | |
GenBank mRNA | BX934896.1 | - | 7 - 8 | 665 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |