Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147517.1 | XP_025003285.1 | 1 - 2 | 112 | 73 | |
GenBank mRNA | BX935016.2 | - | 1 - 2 | 102 | 73 | |
BX935434.1 | - | 1 - 2 | 145 | 73 | ||
CR352812.1 | - | 1 - 2 | 155 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |