Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641618.4 | XP_003641666.1 | 4 - 5 | 578 | 117 | |
XM_004942896.2 | XP_004942953.2 | 4 - 5 | 606 | 117 | ||
XM_025152428.1 | XP_025008196.1 | 4 - 5 | 663 | 117 | ||
XM_025152429.1 | XP_025008197.1 | 4 - 5 | 765 | 117 | ||
XM_422085.5 | XP_422085.1 | 4 - 5 | 658 | 117 | ||
GenBank mRNA | BX932307.2 | - | 4 - 5 | 632 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |