Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199525.1 | NP_001186454.1 | 4 - 5 | 708 | 83 | |
GenBank mRNA | CR354049.1 | - | 4 - 5 | 614 | 83 | |
CR354102.1 | - | 4 - 5 | 701 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |