Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291238.2 | XP_015146724.1 | 3 - 4 | 506 | 119 | |
XM_015291240.2 | XP_015146726.1 | 4 - 5 | 662 | 119 | ||
XM_015291244.2 | XP_015146730.1 | 4 - 5 | 943 | 119 | ||
XM_015291253.2 | XP_015146739.1 | 3 - 4 | 477 | 119 | ||
XM_015291255.2 | XP_015146741.1 | 3 - 4 | 569 | 119 | ||
XM_025142204.1 | XP_024997972.1 | 4 - 5 | 601 | 119 | ||
XM_416454.6 | XP_416454.1 | 3 - 4 | 681 | 119 | ||
GenBank mRNA | CR386658.1 | - | 3 - 4 | 527 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |