Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR387041.1 | - | 1 - 2 | 462 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |