Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277568.1 | NP_001264497.1 | 5 - 6 | 523 | 67 | |
GenBank mRNA | BX932080.1 | - | 4 - 5 | 496 | 67 | |
BX934369.2 | - | 4 - 5 | 493 | 67 | ||
BX936148.1 | - | 4 - 5 | 493 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |