Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277897.2 | XP_015133383.1 | 6 - 7 | 1403 | 83 | |
XM_015277898.2 | XP_015133384.1 | 6 - 7 | 791 | 83 | ||
XM_015277899.2 | XP_015133385.1 | 7 - 8 | 929 | 83 | ||
XM_025146715.1 | XP_025002483.1 | 6 - 7 | 977 | 83 | ||
GenBank mRNA | BX932972.1 | - | 4 - 5 | 472 | 83 | |
CR386062.1 | - | 3 - 4 | 328 | 83 | ||
CR523518.1 | - | 6 - 7 | 743 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |