Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416707.6 | XP_416707.3 | 15 - 16 | 7345 | 136 | |
GenBank mRNA | CR386728.1 | - | 1 - 2 | 53 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |