Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006214.2 | NP_001006214.1 | 2 - 3 | 77 | ||
GenBank mRNA | AJ719994.1 | - | 2 - 3 | 220 | 77 | |
CR388669.1 | - | 2 - 3 | 188 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |