Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291617.2 | XP_015147103.2 | 2 - 3 | 177 | 140 | |
XM_025153641.1 | XP_025009409.1 | 2 - 3 | 1899 | 140 | ||
XM_025153642.1 | XP_025009410.1 | 2 - 3 | 1891 | 140 | ||
GenBank mRNA | CR390566.1 | - | 2 - 3 | 140 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |