Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941226.3 | XP_004941283.1 | 4 | 1525 | 95 | |
XM_015276370.1 | XP_015131856.1 | 4 | 1225 | 95 | ||
XM_015276371.2 | XP_015131857.1 | 4 | 1260 | 95 | ||
XM_015276372.2 | XP_015131858.1 | 6 | 1436 | 95 | ||
XM_025150192.1 | XP_025005960.1 | 5 | 1320 | 95 | ||
GenBank mRNA | BX929915.1 | - | 1 | 626 | 95 | |
CR406270.1 | - | 1 | 552 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |