Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167683.1 | NP_001161155.1 | 5 - 6 | 827 | 102 | |
XM_025149449.1 | XP_025005217.1 | 4 - 5 | 659 | 102 | ||
GenBank mRNA | M58514.1 | - | 5 - 6 | 753 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |