Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198600.1 | NP_001185529.1 | 1 - 2 | 146 | 95 | |
XM_015275703.2 | XP_015131189.1 | 2 - 3 | 261 | 95 | ||
GenBank mRNA | AJ295847.1 | - | 1 - 2 | 146 | 95 | |
BX930971.2 | - | 1 - 2 | 130 | 95 | ||
BX931005.1 | - | 1 - 2 | 62 | 95 | ||
BX934429.1 | - | 1 - 2 | 93 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |