Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP7B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147742.1 | XP_025003510.1 | 1 - 2 | 560 | 73 | |
GenBank mRNA | AY700579.1 | - | 1 - 2 | 83 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |