Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277623.1 | NP_001264552.1 | 1 - 2 | 122 | 139 | |
NM_001277624.1 | NP_001264553.1 | 1 - 2 | 122 | 139 | ||
GenBank mRNA | BG709792.1 | - | 1 - 2 | 122 | 139 | |
BU136195.1 | - | 1 - 2 | 113 | 139 | ||
BU357038.1 | - | 1 - 2 | 116 | 139 | ||
BX929638.1 | - | 1 - 2 | 118 | 139 | ||
BX932269.1 | - | 1 - 2 | 116 | 139 | ||
BX935499.1 | - | 1 - 2 | 113 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |