Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199557.1 | NP_001186486.1 | 3 - 4 | 432 | 89 | |
GenBank mRNA | BX931979.2 | - | 2 - 3 | 255 | 89 | |
BX932028.2 | - | 3 - 4 | 432 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |