Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944615.3 | XP_004944672.1 | 3 - 4 | 678 | 64 | |
XM_414385.6 | XP_414385.2 | 3 - 4 | 865 | 64 | ||
GenBank mRNA | CR386193.1 | - | 2 - 3 | 577 | 64 | |
CR405782.1 | - | 1 - 2 | 291 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |