Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199395.1 | NP_001186324.1 | 4 - 5 | 1376 | 146 | |
GenBank mRNA | CR406449.1 | - | 4 - 5 | 540 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |