Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422088.5 | XP_422088.1 | 1 - 2 | 151 | 67 | |
GenBank mRNA | BX930757.1 | - | 1 - 2 | 130 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |