Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277923.1 | NP_001264852.1 | 6 - 7 | 628 | 94 | |
GenBank mRNA | BX931455.1 | - | 6 - 7 | 628 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |