Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420582.6 | XP_420582.1 | 3 - 4 | 351 | 82 | |
GenBank mRNA | BX932415.1 | - | 1 - 2 | 77 | 82 | |
BX934777.1 | - | 2 - 3 | 273 | 82 | ||
CR385373.1 | - | 3 - 4 | 298 | 82 |
Gene Symbol: SRP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149648.1 | XP_025005416.1 | 2 - 3 | 1001 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |