Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288856.2 | XP_015144342.1 | 2 - 3 | 209 | 93 | |
GenBank mRNA | BX934634.1 | - | 2 - 3 | 202 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |