Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277660.1 | NP_001264589.1 | 6 - 7 | 893 | 114 | |
XM_015278303.2 | XP_015133789.2 | 7 - 8 | 925 | 114 | ||
XM_015278304.2 | XP_015133790.1 | 7 - 8 | 1041 | 114 | ||
XM_015278305.2 | XP_015133791.1 | 7 - 8 | 1028 | 114 | ||
XM_015278306.2 | XP_015133792.1 | 6 - 7 | 959 | 114 | ||
XM_015278307.2 | XP_015133793.1 | 7 - 8 | 1007 | 114 | ||
XM_015278308.2 | XP_015133794.1 | 7 - 8 | 909 | 114 | ||
XM_025149575.1 | XP_025005343.1 | 7 - 8 | 998 | 114 | ||
XM_025149576.1 | XP_025005344.1 | 8 - 9 | 997 | 114 | ||
XM_025149577.1 | XP_025005345.1 | 8 - 9 | 1012 | 114 | ||
XM_025149578.1 | XP_025005346.1 | 6 - 7 | 899 | 114 | ||
GenBank mRNA | BX934826.1 | - | 6 - 7 | 885 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |