Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414755.5 | XP_414755.2 | 6 - 7 | 1440 | 72 | |
GenBank mRNA | BX934565.2 | - | 4 - 5 | 602 | 72 | |
BX936040.1 | - | 4 - 5 | 610 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |