Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004940932.3 | XP_004940989.1 | 5 - 6 | 830 | 91 | |
XM_015285369.2 | XP_015140855.1 | 5 - 6 | 648 | 91 | ||
XM_015285370.2 | XP_015140856.1 | 5 - 6 | 920 | 91 | ||
XM_025150106.1 | XP_025005874.1 | 4 - 5 | 546 | 91 | ||
XM_420403.6 | XP_420403.1 | 5 - 6 | 612 | 91 | ||
GenBank mRNA | BX950414.1 | - | 5 - 6 | 567 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |