Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277734.1 | NP_001264663.1 | 1 - 2 | 90 | 61 | |
GenBank mRNA | CR353735.1 | - | 1 - 2 | 76 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |