Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416468.6 | XP_416468.3 | 3 - 4 | 1176 | 90 | |
GenBank mRNA | BX930002.2 | - | 3 - 4 | 539 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |