Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935651.3 | XP_004935708.1 | 2 - 3 | 243 | 89 | |
XM_419691.6 | XP_419691.2 | 2 - 3 | 254 | 89 | ||
GenBank mRNA | BX929385.2 | - | 2 - 3 | 239 | 89 | |
BX932702.1 | - | 2 - 3 | 228 | 89 | ||
BX935794.2 | - | 2 - 3 | 225 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |