Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277886.1 | NP_001264815.1 | 7 - 8 | 633 | 109 | |
XM_015278223.2 | XP_015133709.1 | 7 - 8 | 648 | 109 | ||
XM_015278224.2 | XP_015133710.1 | 7 - 8 | 649 | 109 | ||
XM_025149547.1 | XP_025005315.1 | 7 - 8 | 649 | 109 | ||
GenBank mRNA | BX930950.1 | - | 7 - 8 | 619 | 109 | |
BX935900.2 | - | 7 - 8 | 635 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |