Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031287.2 | NP_001026458.2 | 3 - 4 | 337 | 76 | |
XM_025152880.1 | XP_025008648.1 | 3 - 4 | 456 | 76 | ||
GenBank mRNA | AJ720201.1 | - | 3 - 4 | 335 | 76 | |
CO420070.1 | - | 3 - 4 | 354 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |