Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012855.3 | NP_001012873.3 | 5 - 6 | 944 | 147 | |
XM_015281400.2 | XP_015136886.1 | 5 - 6 | 1411 | 147 | ||
GenBank mRNA | AJ721135.1 | - | 5 - 6 | 613 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |