Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938073.3 | XP_004938130.1 | 4 | 89 | ||
XM_004938075.3 | XP_004938132.1 | 3 | 89 | |||
XM_004938076.3 | XP_004938133.1 | 3 | 89 | |||
GenBank mRNA | AY646851.1 | - | 1 | 56 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |