Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277607.1 | NP_001264536.1 | 4 - 5 | 673 | 87 | |
NM_001277608.1 | NP_001264537.1 | 5 - 6 | 467 | 87 | ||
NM_001277609.1 | NP_001264538.1 | 4 - 5 | 643 | 87 | ||
NM_001277610.1 | NP_001264539.1 | 5 - 6 | 437 | 87 | ||
GenBank mRNA | BX929369.1 | - | 5 - 6 | 425 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |