Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947639.3 | XP_004947696.1 | 7 | 2453 | 91 | |
XM_004947640.3 | XP_004947697.1 | 8 | 2062 | 91 | ||
XM_004947641.3 | XP_004947698.1 | 8 | 2035 | 91 | ||
XM_015297496.2 | XP_015152982.1 | 7 | 2424 | 91 | ||
XM_025142841.1 | XP_024998609.1 | 8 | 2197 | 91 | ||
XM_025142842.1 | XP_024998610.1 | 7 | 2318 | 91 | ||
XM_025142843.1 | XP_024998611.1 | 7 | 2455 | 91 | ||
XM_417690.6 | XP_417690.3 | 7 | 2291 | 91 | ||
GenBank mRNA | CR388785.1 | - | 1 | 849 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |