Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943514.3 | XP_004943571.1 | 2 | 2746 | 105 | |
XM_015291773.2 | XP_015147259.1 | 5 | 4445 | 105 | ||
XM_015291775.2 | XP_015147261.1 | 6 | 4550 | 105 | ||
XM_025153491.1 | XP_025009259.1 | 5 | 5096 | 105 | ||
XM_025153492.1 | XP_025009260.1 | 4 | 5006 | 105 | ||
XM_025153493.1 | XP_025009261.1 | 4 | 7790 | 105 | ||
XM_025153494.1 | XP_025009262.1 | 7 | 4671 | 105 | ||
XM_025153495.1 | XP_025009263.1 | 6 | 4529 | 105 | ||
XM_025153496.1 | XP_025009264.1 | 4 | 7854 | 105 | ||
XM_025153497.1 | XP_025009265.1 | 7 | 4735 | 105 | ||
XM_025153498.1 | XP_025009266.1 | 5 | 4236 | 105 | ||
XM_025153499.1 | XP_025009267.1 | 6 | 4593 | 105 | ||
XM_025153500.1 | XP_025009268.1 | 3 | 2813 | 105 | ||
GenBank mRNA | CR405896.1 | - | 1 | 2954 | 105 |
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025153490.1 | XP_025009258.1 | 4 | 8928 | 105 | |
XM_426717.5 | XP_426717.2 | 4 | 9454 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |