Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282331.1 | NP_001269260.1 | 1 - 2 | 98 | ||
GenBank mRNA | BU382037.1 | - | 1 - 2 | 114 | 98 | |
CD217667.1 | - | 1 - 2 | 98 | |||
CR406781.1 | - | 1 - 2 | 115 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |