Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278024.1 | NP_001264953.1 | 1 - 2 | 281 | 120 | |
GenBank mRNA | AJ720615.1 | - | 1 - 2 | 281 | 120 | |
BX934705.1 | - | 1 - 2 | 174 | 120 | ||
CR353363.1 | - | 1 - 2 | 169 | 120 | ||
CR407495.1 | - | 1 - 2 | 150 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |