Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277356.1 | NP_001264285.1 | 5 - 6 | 505 | 61 | |
NM_001277357.1 | NP_001264286.1 | 5 - 6 | 465 | 61 | ||
XM_025147649.1 | XP_025003417.1 | 5 - 6 | 612 | 61 | ||
GenBank mRNA | AW198506.1 | - | 1 - 2 | 121 | 61 | |
BX930706.1 | - | 5 - 6 | 502 | 61 | ||
BX931538.1 | - | 5 - 6 | 505 | 61 | ||
CF253209.1 | - | 5 - 6 | 467 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |