Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284385.2 | XP_015139871.1 | 2 - 3 | 2642 | 87 | |
GenBank mRNA | BX931270.1 | - | 1 - 2 | 234 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |