Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277337.1 | NP_001264266.1 | 5 - 6 | 615 | 113 | |
XM_015278680.2 | XP_015134166.1 | 5 - 6 | 474 | 113 | ||
GenBank mRNA | BX934609.1 | - | 5 - 6 | 493 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |