Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199668.1 | NP_001186597.1 | 4 - 5 | 678 | 76 | |
XM_015287078.2 | XP_015142564.1 | 4 - 5 | 777 | 76 | ||
XM_015287079.2 | XP_015142565.1 | 4 - 5 | 668 | 76 | ||
GenBank mRNA | BX934990.1 | - | 4 - 5 | 619 | 76 | |
CN228613.1 | - | 4 - 5 | 597 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |