Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR353653.1 | - | 1 - 2 | 231 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |